Assay Details
| Gene Symbol: Ecel1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277925.1 | NP_001264854.1 | 5 - 6 | 1276 | 81 | |
| NM_021306.3 | NP_067281.2 | 5 - 6 | 1268 | 81 | ||
| GenBank mRNA | AB026294.1 | - | 4 - 5 | 1096 | 81 | |
| AK039467.1 | - | 5 - 6 | 82 | |||
| AK159365.1 | - | 5 - 6 | 1278 | 81 | ||
| BC057569.1 | - | 4 - 5 | 1098 | 6413731 | 81 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ecel1
Genomic Map