Assay Details
| Gene Symbol: Gm9530 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK029591.1 | - | 1 - 2 | 2077 | 70 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
calcium channel, voltage-dependent, R type, alpha 1E subunit |
Cacna1e |
A430040I15, BII, Cach6, Cacnl1a6, Cav2.3, Cchra1, alpha1E |
-195.9kb | Sense | Chr. 1 - 154390806 - 154884782 on NCBI Build GRCm38 |
| Downstream | immediate early response 5 | Ier5 | 13.1kb | Sense | Chr. 1 - 155096367 - 155099636 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Gm9530
Genomic Map