Assay Details
| Gene Symbol: Hils1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018792.1 | NP_061262.1 | 1 | 742 | 81 | |
| GenBank mRNA | AB022320.1 | - | 1 | 742 | 81 | |
| AB045724.1 | - | 1 | 735 | 81 | ||
| AK005718.1 | - | 1 | 773 | 81 | ||
| AY286319.1 | - | 1 | 742 | 81 | ||
| BC050966.1 | - | 1 | 775 | 81 | ||
| BC061119.1 | - | 1 | 805 | 6773493 | 81 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Hils1
Genomic Map