Assay Details
| Gene Symbol: Pdk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133667.2 | NP_598428.2 | 9 - 10 | 1087 | 109 | |
| GenBank mRNA | AF267660.1 | - | 9 - 10 | 1048 | 109 | |
| AK002647.1 | - | 9 - 10 | 1031 | 109 | ||
| BC021764.1 | - | 9 - 10 | 1041 | 3488813 | 109 | |
| CK618575.1 | - | 2 - 3 | 142 | 109 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pdk2
Genomic Map