Assay Details
| Gene Symbol: Lonrf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028894.1 | NP_083170.1 | 8 - 9 | 1964 | 87 | |
| GenBank mRNA | AK016522.1 | - | 8 - 9 | 1964 | 87 | |
| AK167341.1 | - | 8 - 9 | 2023 | 87 | ||
| AK185175.1 | - | 1 - 2 | 57 | 87 | ||
| BC120688.1 | - | 8 - 9 | 1866 | 40129611 | 87 | |
| BC120690.1 | - | 8 - 9 | 1866 | 40129613 | 87 | |
More Information
Updated Mapping Information
Set Membership: |
|

Lonrf3
Genomic Map