Assay Details
| Gene Symbol: Bhlhb9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198161.2 | NP_937804.1 | 2 - 3 | 330 | 143 | |
| GenBank mRNA | AK143861.1 | - | 2 - 3 | 253 | 143 | |
| AK157023.1 | - | 2 - 3 | 284 | 143 | ||
| CK625652.1 | - | 2 - 3 | 319 | 143 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK143861.1, AK157023.1, NM_198161.2 transcripts.
Set Membership: |
|

Bhlhb9
Genomic Map