Assay Details
| Gene Symbol: Slc11a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001146161.1 | NP_001139633.1 | 12 - 13 | 1328 | 61 | |
| NM_008732.2 | NP_032758.2 | 12 - 13 | 1328 | 61 | ||
| GenBank mRNA | AF029758.1 | - | 12 - 13 | 1263 | 61 | |
| AK049856.1 | - | 12 - 13 | 1276 | 61 | ||
| AK083478.1 | - | 12 - 13 | 1274 | 61 | ||
| AK148276.1 | - | 12 - 13 | 1276 | 61 | ||
| AK193963.1 | - | 1 - 2 | 47 | 61 | ||
| AK205883.1 | - | 1 - 2 | 47 | 61 | ||
| BC019137.1 | - | 12 - 13 | 1302 | 5040976 | 61 | |
| L33415.1 | - | 12 - 13 | 1256 | 61 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Slc11a2
Genomic Map