Assay Details
| Gene Symbol: Slc11a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001146161.1 | NP_001139633.1 | 1 - 2 | 91 | 59 | |
| NM_008732.2 | NP_032758.2 | 1 - 2 | 91 | 59 | ||
| GenBank mRNA | AK049856.1 | - | 1 - 2 | 39 | 59 | |
| AK083478.1 | - | 1 - 2 | 37 | 59 | ||
| BC019137.1 | - | 1 - 2 | 65 | 5040976 | 59 | |
| CJ057283.1 | - | 1 - 2 | 92 | 59 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK049856.1, AK083478.1, BC019137.1, NM_001146161.1, NM_008732.2 transcripts.
Set Membership: |
|

Slc11a2
Genomic Map