Assay Details
| Gene Symbol: Dennd2c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177857.1 | NP_808525.2 | 11 - 12 | 2074 | 75 | |
| GenBank mRNA | AK044389.1 | - | 10 - 11 | 2450 | 75 | |
| AK137637.1 | - | 6 - 7 | 681 | 75 | ||
| AK172232.1 | - | 7 - 8 | 796 | 75 | ||
| BC046440.1 | - | 7 - 8 | 542 | 75 | ||
| BC060666.1 | - | 11 - 12 | 2074 | 6841982 | 75 | |
| BC066182.1 | - | 4 - 5 | 320 | 75 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Dennd2c
Genomic Map