Assay Details
| Gene Symbol: Dennd2c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177857.1 | NP_808525.2 | 12 - 13 | 2213 | 68 | |
| GenBank mRNA | AK044389.1 | - | 11 - 12 | 2589 | 68 | |
| AK137637.1 | - | 7 - 8 | 820 | 68 | ||
| AK172232.1 | - | 8 - 9 | 935 | 68 | ||
| BC046440.1 | - | 8 - 9 | 681 | 68 | ||
| BC060666.1 | - | 12 - 13 | 2213 | 6841982 | 68 | |
| BC066182.1 | - | 5 - 6 | 459 | 68 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Dennd2c
Genomic Map