Assay Details
| Gene Symbol: Dennd2c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177857.1 | NP_808525.2 | 6 - 7 | 1587 | 138 | |
| GenBank mRNA | AK044389.1 | - | 5 - 6 | 1963 | 138 | |
| BC060666.1 | - | 6 - 7 | 1587 | 6841982 | 138 | |
More Information
Set Membership: |
|

Dennd2c
Genomic Map