Assay Details
| Gene Symbol: Dennd2c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177857.1 | NP_808525.2 | 7 - 8 | 1632 | 96 | |
| GenBank mRNA | AK044389.1 | - | 6 - 7 | 2008 | 96 | |
| BC046440.1 | - | 2 - 3 | 100 | 96 | ||
| BC060666.1 | - | 7 - 8 | 1632 | 6841982 | 96 | |
More Information
Set Membership: |
|

Dennd2c
Genomic Map