Assay Details
| Gene Symbol: Dennd2c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177857.1 | NP_808525.2 | 8 - 9 | 1813 | 73 | |
| GenBank mRNA | AK044389.1 | - | 7 - 8 | 2189 | 73 | |
| AK137637.1 | - | 3 - 4 | 420 | 73 | ||
| AK172232.1 | - | 4 - 5 | 535 | 73 | ||
| BC046440.1 | - | 3 - 4 | 281 | 73 | ||
| BC060666.1 | - | 8 - 9 | 1813 | 6841982 | 73 | |
| BC066182.1 | - | 1 - 2 | 59 | 73 | ||
More Information
Set Membership: |
|

Dennd2c
Genomic Map