Assay Details
| Gene Symbol: Hsd17b3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008291.3 | NP_032317.2 | 6 - 7 | 554 | 74 | |
| GenBank mRNA | U66827.1 | - | 6 - 7 | 539 | 74 | |
More Information
Updated Mapping Information
Set Membership: |
|

Hsd17b3
Genomic Map