Assay Details
| Gene Symbol: Nup62cl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081668.1 | NP_001075137.1 | 7 - 8 | 870 | 77 | |
| GenBank mRNA | BC100367.1 | - | 7 - 8 | 854 | 77 | |
More Information
Updated Mapping Information
Set Membership: |
|

Nup62cl
Genomic Map