| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0700001898.mm.1 |
TC0700000257.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700001898.mm.1 |
NM_028480.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700001898.mm.1 |
NM_028480.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700001893.mm.1 |
TC0700000257.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700001893.mm.1 |
NM_028480.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700001893.mm.1 |
NM_028480.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700001896.mm.1 |
TC0700000257.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700001896.mm.1 |
NM_028480.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700001896.mm.1 |
NM_028480.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700001899.mm.1 |
TC0700000257.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700001899.mm.1 |
NM_028480.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700001899.mm.1 |
NM_028480.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700001894.mm.1 |
TC0700000257.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700001894.mm.1 |
NM_028480.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700001894.mm.1 |
NM_028480.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700001895.mm.1 |
TC0700000257.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700001895.mm.1 |
NM_028480.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700001895.mm.1 |
NM_028480.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700001897.mm.1 |
TC0700000257.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700001897.mm.1 |
NM_028480.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700001897.mm.1 |
NM_028480.2 |
Refseq |