Assay Details
| Gene Symbol: Krt32 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001159374.2 | NP_001152846.2 | 4 - 5 | 882 | 59 | |
| GenBank mRNA | AK009595.1 | - | 2 - 3 | 209 | 59 | |
| BC117553.1 | - | 4 - 5 | 772 | 40104816 | 59 | |
| BC118064.1 | - | 4 - 5 | 772 | 40104817 | 59 | |
| X75649.1 | - | 4 - 5 | 772 | 59 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Krt32
Genomic Map