Assay Details
| Gene Symbol: Nt5c3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102650.1 | NP_001096120.1 | 3 - 4 | 214 | 100 | |
| NM_026561.4 | NP_080837.3 | 3 - 4 | 214 | 100 | ||
| GenBank mRNA | AK006972.1 | - | 4 - 5 | 423 | 100 | |
| AK082816.1 | - | 3 - 4 | 200 | 100 | ||
| AK148222.1 | - | 3 - 4 | 190 | 100 | ||
| AK162770.1 | - | 2 - 3 | 292 | 100 | ||
| AK166393.1 | - | 3 - 4 | 190 | 100 | ||
| AK211190.1 | - | 1 - 2 | 78 | 100 | ||
| BC015307.1 | - | 2 - 3 | 169 | 4239117 | 100 | |
| BY047327.1 | - | 3 - 4 | 214 | 100 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK006972.1, AK082816.1, AK162770.1, BC015307.1 transcripts.
Set Membership: |
|

Nt5c3b
Genomic Map