Assay Details
| Gene Symbol: Gm11595 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001126322.1 | NP_001119794.1 | 1 | 944 | 68 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1963573 | NM_001126322.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2153126 | NM_001126322.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1963576 | NM_001126322.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1963576 | NM_001126322.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2153126 | NM_001126322.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1963573 | NM_001126322.1 | mouse_transcriptome |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28064350 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28064270 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28064287 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28064260 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28064289 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28064335 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28064311 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28064257 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28064266 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28064248 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28064303 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28064284 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28064268 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28064339 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28064276 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28064321 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28064342 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28064262 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28064349 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28064264 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28064338 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28064258 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28064272 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28064336 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28064304 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28064344 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28064285 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28064347 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28064253 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28064250 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28064278 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08991299 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28064268 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28064316 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28064284 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28064325 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28064275 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28064272 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28064309 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28064304 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28064319 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28064280 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28064287 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28064276 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28064270 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28064317 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28064311 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28064260 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28064303 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09455557 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28064285 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28064328 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28064258 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28064306 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28064321 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28064271 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09547932 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28064281 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28064262 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28064266 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09280390 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28064289 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09547950 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28064314 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10390935 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10390948 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10390943 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10390934 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 393843 | - | - |

Gm11595
Genomic Map