Assay Details
| Gene Symbol: Fcmr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026976.2 | NP_081252.1 | 6 - 7 | 1115 | 130 | |
| GenBank mRNA | AK007714.1 | - | 6 - 7 | 1116 | 130 | |
| AK155943.1 | - | 6 - 7 | 1116 | 130 | ||
| AY227385.1 | - | 6 - 7 | 1032 | 130 | ||
| AY227386.1 | - | 6 - 7 | 1032 | 130 | ||
| AY227387.1 | - | 6 - 7 | 1032 | 130 | ||
| AY227388.1 | - | 6 - 7 | 1032 | 130 | ||
| AY227389.1 | - | 6 - 7 | 1032 | 130 | ||
| AY227390.1 | - | 6 - 7 | 1032 | 130 | ||
| AY227391.1 | - | 6 - 7 | 1032 | 130 | ||
| AY227392.1 | - | 6 - 7 | 1032 | 130 | ||
| AY227393.1 | - | 6 - 7 | 1032 | 130 | ||
| BC116674.1 | - | 6 - 7 | 1220 | 40106407 | 130 | |
| BC118505.1 | - | 6 - 7 | 1220 | 40106408 | 130 | |
More Information
Updated Mapping Information
Set Membership: |
|

Fcmr
Genomic Map