Assay Details
| Gene Symbol: Fcmr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026976.2 | NP_081252.1 | 7 - 8 | 1175 | 106 | |
| GenBank mRNA | AK007714.1 | - | 7 - 8 | 1176 | 106 | |
| AK155943.1 | - | 7 - 8 | 1176 | 106 | ||
| AY227385.1 | - | 7 - 8 | 1092 | 106 | ||
| AY227386.1 | - | 7 - 8 | 1092 | 106 | ||
| AY227387.1 | - | 7 - 8 | 1092 | 106 | ||
| AY227388.1 | - | 7 - 8 | 1092 | 106 | ||
| AY227389.1 | - | 7 - 8 | 1092 | 106 | ||
| AY227390.1 | - | 7 - 8 | 1092 | 106 | ||
| AY227391.1 | - | 7 - 8 | 1092 | 106 | ||
| AY227392.1 | - | 7 - 8 | 1092 | 106 | ||
| AY227393.1 | - | 7 - 8 | 1092 | 106 | ||
| BC116674.1 | - | 7 - 8 | 1280 | 40106407 | 106 | |
| BC118505.1 | - | 7 - 8 | 1280 | 40106408 | 106 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20612317 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20612316 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20612314 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20612315 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P378298 | AK007714.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P378298 | NM_026976.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P378298 | AK007714.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P378298 | NM_026976.2 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 118382 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 699181 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 777923 | AK086423.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 777923 | AK142756.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 9383 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 81207 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 760988 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 497818 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 358651 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 740000 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 82135 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 105134 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 163118 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P135403 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P153840 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P129930 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P378298 | AK007714.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P378298 | NM_026976.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P378298 | AK007714.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P378298 | NM_026976.2 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20612246 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20612240 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20612230 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20612252 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20612228 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20612248 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20612253 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20612222 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20612236 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20612241 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20612226 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20612244 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20612249 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20612237 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20612238 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20612229 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20612232 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20612224 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20612233 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20612235 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20612250 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P04262861 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P04262848 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P153840 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P129930 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P04262828 | - | - |
| Whole Mouse Genome G4122A | A_51_P378298 | AK007714.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P378298 | NM_026976.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P378298 | AK007714.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P378298 | NM_026976.2 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P153840 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P129930 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01095290 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01095294 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00167165 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00167169 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01095303 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00167176 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20612317 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20612316 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20612314 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20612315 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20612397 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20612226 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20612308 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20612258 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20612361 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20612406 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20612249 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20612267 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20612221 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20612278 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20612230 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20612316 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20612311 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20612299 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20612331 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20612306 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20612403 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20612304 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20612215 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20612292 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20612266 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20612332 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20612232 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20612340 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20612242 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20612372 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20612401 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20612373 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20612326 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20612377 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20612354 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20612353 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20612367 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20612320 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20612400 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20612216 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20612237 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20612244 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20612383 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20612224 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20612356 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20612235 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20612288 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20612374 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20612256 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20612365 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20612376 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20612295 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20612212 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20612333 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20612368 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20612250 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20612269 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20612349 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20612218 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20612248 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20612302 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20612335 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20612408 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20612328 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20612386 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20612290 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20612324 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20612246 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20612390 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20612363 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20612357 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20612351 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20612344 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20612330 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20612346 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20612238 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20612310 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20612379 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20612275 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20612338 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20612272 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20612236 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20612271 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20612325 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20612213 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20612208 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20612318 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20612348 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20612291 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20612395 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20612381 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20612261 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20612297 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20612276 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20612264 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20612285 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20612385 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20612222 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20612281 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20612241 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20612410 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20612387 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20612370 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20612280 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20612252 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20612249 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00805851 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20612216 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20612331 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20612267 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20612230 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00167169 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20612233 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20612290 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20612218 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20612338 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20612299 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00330895 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20612264 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20612259 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20612323 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20612302 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20612226 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20612220 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20612232 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20612340 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20612248 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20612250 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00618347 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20612288 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20612287 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00525155 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20612332 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20612254 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20612324 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20612335 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20612271 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20612235 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20612260 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20612228 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20612295 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20612269 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20612275 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20612263 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20612320 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20612237 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20612236 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00211776 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20612325 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00618348 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20612285 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20612221 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20612253 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20612215 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20612292 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00972532 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20612262 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20612229 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20612261 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20612297 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20612244 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20612238 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20612252 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20612326 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00525157 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20612328 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20612241 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20612291 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20612318 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20612333 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00972531 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20612214 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20612242 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10349600 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10349599 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10349596 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10349594 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10349602 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10349595 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10349597 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10349601 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10349598 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1429889_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10349593 | - | - |
| Mouse Genome 430 2.0 Array | 1429889_at | - | - |
| Mouse Genome 430 2.0 Array | 1429889_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 69169_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 69169_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0100009377.mm.1 | NM_026976.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100009377.mm.1 | NM_026976.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100009377.mm.1 | TC0100001166.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1429889_PM_at | - | - |
| Mouse Expression Set 430 | 1429889_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1429889_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1429889_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 836319 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1221820 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1221820 | - | - |

Fcmr
Genomic Map