Assay Details
| Gene Symbol: Dusp14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK166840.1 | - | 1 - 2 | 82 | 73 | |
| BC052836.1 | - | 1 - 2 | 106 | 30049007 | 73 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK166840.1, BC052836.1 transcripts.
Set Membership: |
|

Dusp14
Genomic Map