Assay Details
| Gene Symbol: Dusp14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019819.3 | NP_062793.2 | 2 - 3 | 177 | 92 | |
| GenBank mRNA | AK079362.1 | - | 2 - 3 | 178 | 92 | |
| AK163738.1 | - | 2 - 3 | 222 | 92 | ||
| AK166230.1 | - | 2 - 3 | 175 | 92 | ||
| AK166840.1 | - | 2 - 3 | 173 | 92 | ||
| BC002130.1 | - | 2 - 3 | 142 | 3592420 | 92 | |
| BC052836.1 | - | 2 - 3 | 197 | 30049007 | 92 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK079362.1, AK166230.1, AK166840.1, BC002130.1, BC052836.1, NM_019819.3 transcripts.
Set Membership: |
|

Dusp14
Genomic Map