Assay Details
| Gene Symbol: Mmp28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_080453.2 | NP_536701.1 | 3 - 4 | 486 | 71 | |
| NM_172797.2 | NP_766385.1 | 3 - 4 | 486 | 71 | ||
| GenBank mRNA | AK051258.1 | - | 3 - 4 | 549 | 71 | |
| AY065653.1 | - | 3 - 4 | 486 | 71 | ||
| AY071828.1 | - | 3 - 4 | 486 | 71 | ||
| AY071829.1 | - | 3 - 4 | 486 | 71 | ||
| BC137921.1 | - | 3 - 4 | 441 | 8860940 | 71 | |
| BC145048.1 | - | 3 - 4 | 441 | 9053596 | 71 | |
| BC145049.1 | - | 3 - 4 | 441 | 9053597 | 71 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mmp28
Genomic Map