Assay Details
| Gene Symbol: Prss32 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027220.2 | NP_081496.2 | 1 - 2 | 151 | 64 | |
| GenBank mRNA | AK008023.1 | - | 1 - 2 | 148 | 64 | |
| AY266139.1 | - | 1 - 2 | 138 | 64 | ||
| BC024903.1 | - | 1 - 2 | 151 | 4208033 | 64 | |
More Information
Updated Mapping Information
Set Membership: |
|

Prss32
Genomic Map