Assay Details
| Gene Symbol: Arhgef6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_152801.2 | NP_690014.2 | 19 - 20 | 2244 | 104 | |
| GenBank mRNA | AF393831.1 | - | 19 - 20 | 2244 | 104 | |
| AK006635.1 | - | 12 - 13 | 1243 | 104 | ||
| BC019723.1 | - | 2 - 3 | 371 | 104 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Arhgef6
Genomic Map