Assay Details
| Gene Symbol: Amdhd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172935.4 | NP_766523.2 | 8 - 9 | 1041 | 61 | |
| GenBank mRNA | AK077584.1 | - | 8 - 9 | 1024 | 61 | |
| AK090009.1 | - | 8 - 9 | 1043 | 61 | ||
| AK154209.1 | - | 8 - 9 | 1042 | 61 | ||
| AK159860.1 | - | 8 - 9 | 1038 | 61 | ||
| AK168432.1 | - | 8 - 9 | 999 | 61 | ||
| AK179853.1 | - | 1 - 2 | 60 | 61 | ||
| AK180149.1 | - | 1 - 2 | 60 | 61 | ||
| AK180231.1 | - | 1 - 2 | 60 | 61 | ||
| AK200617.1 | - | 3 - 4 | 360 | 61 | ||
| BC037005.1 | - | 8 - 9 | 1077 | 5101453 | 61 | |
More Information
Updated Mapping Information
Set Membership: |
|

Amdhd2
Genomic Map