Assay Details
| Gene Symbol: Unkl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001197024.1 | NP_001183953.1 | 3 - 4 | 485 | 59 | |
| NM_001290736.1 | NP_001277665.1 | 3 - 4 | 496 | 59 | ||
| GenBank mRNA | BC059910.1 | - | 3 - 4 | 496 | 59 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001290736.1 transcripts.
Set Membership: |
|

Unkl
Genomic Map