| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1700002982.mm.1 |
NM_001079929.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700002982.mm.1 |
TC1700000363.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700002995.mm.1 |
NM_001079929.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700002995.mm.1 |
TC1700000363.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700002988.mm.1 |
NM_001079929.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700002988.mm.1 |
TC1700000363.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700002989.mm.1 |
NM_001079929.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700002989.mm.1 |
TC1700000363.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700002981.mm.1 |
NM_001079929.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700002981.mm.1 |
TC1700000363.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700002983.mm.1 |
NM_001079929.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700002983.mm.1 |
TC1700000363.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700002984.mm.1 |
NM_001079929.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700002984.mm.1 |
TC1700000363.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700002996.mm.1 |
NM_001079929.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700002996.mm.1 |
TC1700000363.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700002978.mm.1 |
NM_001079929.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700002978.mm.1 |
TC1700000363.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700002980.mm.1 |
NM_001079929.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700002980.mm.1 |
TC1700000363.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700002985.mm.1 |
NM_001079929.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700002985.mm.1 |
TC1700000363.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700002994.mm.1 |
NM_001079929.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700002994.mm.1 |
TC1700000363.mm.1 |
MTA TC |