Assay Details
| Gene Symbol: Ccdc154 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079929.2 | NP_001073398.2 | 12 - 13 | 1780 | 95 | |
| GenBank mRNA | AK133084.1 | - | 13 - 14 | 1897 | 95 | |
| JN935900.1 | - | 11 - 12 | 1399 | 95 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ccdc154
Genomic Map