Assay Details
| Gene Symbol: Clcn7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011930.3 | NP_036060.1 | 10 - 11 | 996 | 63 | |
| GenBank mRNA | AK030444.1 | - | 10 - 11 | 997 | 63 | |
| AK128981.1 | - | 10 - 11 | 964 | 63 | ||
| AK150395.1 | - | 10 - 11 | 907 | 63 | ||
| AK191479.1 | - | 1 - 2 | 64 | 63 | ||
| AK201127.1 | - | 1 - 2 | 64 | 63 | ||
| AK204909.1 | - | 1 - 2 | 64 | 63 | ||
| AK206010.1 | - | 1 - 2 | 64 | 63 | ||
| AK207109.1 | - | 1 - 2 | 42 | 63 | ||
| AK214309.1 | - | 1 - 2 | 64 | 63 | ||
| AK215539.1 | - | 1 - 2 | 64 | 63 | ||
| BC050907.1 | - | 10 - 11 | 968 | 5698121 | 63 | |
| BC053049.1 | - | 10 - 11 | 968 | 5698121 | 63 | |
| BC054799.1 | - | 10 - 11 | 976 | 6849205 | 63 | |
More Information
Updated Mapping Information
Set Membership: |
|

Clcn7
Genomic Map