Assay Details
| Gene Symbol: Pigh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029988.2 | NP_084264.1 | 2 - 3 | 442 | 91 | |
| GenBank mRNA | AK030744.1 | - | 2 - 3 | 406 | 91 | |
| AK167892.1 | - | 2 - 3 | 424 | 91 | ||
| BC086806.1 | - | 2 - 3 | 393 | 30603877 | 91 | |
| BY737882.1 | - | 2 - 3 | 424 | 91 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pigh
Genomic Map