Assay Details
| Gene Symbol: Vti1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016800.3 | NP_058080.2 | 4 - 5 | 667 | 67 | |
| GenBank mRNA | AF035208.1 | - | 4 - 5 | 554 | 67 | |
| AF035821.1 | - | 4 - 5 | 540 | 67 | ||
| AK160624.1 | - | 3 - 4 | 601 | 67 | ||
| BC002290.1 | - | 3 - 4 | 592 | 67 | ||
| BC009810.1 | - | 4 - 5 | 597 | 4220215 | 67 | |
| BC080204.1 | - | 4 - 5 | 643 | 3672445 | 67 | |
| CX239415.1 | - | 4 - 5 | 644 | 67 | ||
More Information
Set Membership: |
|

Vti1b
Genomic Map