Assay Details
| Gene Symbol: Plekhd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001177503.1 | NP_001170974.1 | 7 - 8 | 931 | 59 | |
| GenBank mRNA | AK028383.1 | - | 7 - 8 | 881 | 59 | |
| BC137602.1 | - | 7 - 8 | 714 | 8860621 | 59 | |
More Information
Updated Mapping Information
Set Membership: |
|

Plekhd1
Genomic Map