Assay Details
| Gene Symbol: Numb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001272055.1 | NP_001258984.1 | 2 - 3 | 216 | 76 | |
| NM_001272056.1 | NP_001258985.1 | 4 - 5 | 446 | 76 | ||
| NR_073563.1 | 5 - 6 | 563 | 76 | |||
| GenBank mRNA | AF169191.1 | - | 2 - 3 | 201 | 76 | |
| AK004553.1 | - | 5 - 6 | 471 | 76 | ||
| BC033459.1 | - | 5 - 6 | 487 | 3991630 | 76 | |
| U70674.1 | - | 4 - 5 | 367 | 76 | ||
More Information
Set Membership: |
|

Numb
Genomic Map