| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1200014113.mm.1 |
NM_001272056.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200014113.mm.1 |
NR_073563.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200014113.mm.1 |
NR_073564.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200014113.mm.1 |
TC1200002053.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1200014122.mm.1 |
NR_073563.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200014122.mm.1 |
NR_073564.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200014122.mm.1 |
TC1200002053.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1200014112.mm.1 |
NM_001272056.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200014112.mm.1 |
NR_073563.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200014112.mm.1 |
NR_073564.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200014112.mm.1 |
TC1200002053.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1200014116.mm.1 |
NR_073563.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200014116.mm.1 |
NR_073564.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200014116.mm.1 |
TC1200002053.mm.1 |
MTA TC |