Assay Details
| Gene Symbol: Dpf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001267625.1 | NP_001254554.1 | 10 - 11 | 1098 | 88 | |
| GenBank mRNA | AK039011.1 | - | 10 - 11 | 1079 | 88 | |
More Information
Updated Mapping Information
Set Membership: |
|

Dpf3
Genomic Map