Assay Details
| Gene Symbol: Dlst | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_030225.4 | NP_084501.1 | 10 - 11 | 835 | 63 | |
| GenBank mRNA | AK019713.2 | - | 832 | 63 | ||
| AK054053.1 | - | 10 - 11 | 789 | 63 | ||
| AK149664.1 | - | 10 - 11 | 811 | 63 | ||
| AK158877.1 | - | 10 - 11 | 836 | 63 | ||
| AK168570.1 | - | 10 - 11 | 811 | 63 | ||
| AK169943.1 | - | 10 - 11 | 789 | 63 | ||
| AK203473.1 | - | 1 - 2 | 38 | 63 | ||
| AK205241.1 | - | 1 - 2 | 38 | 63 | ||
| AK214016.1 | - | 1 - 2 | 92 | 63 | ||
| AK220022.1 | - | 1 - 2 | 38 | 63 | ||
| BC006702.1 | - | 10 - 11 | 772 | 3583936 | 63 | |
| CT010197.1 | - | 10 - 11 | 771 | 63 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Dlst
Genomic Map