Assay Details
| Gene Symbol: Jdp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_030887.2 | NP_112149.2 | 1 - 2 | 202 | 103 | |
| GenBank mRNA | AB077438.1 | - | 1 - 2 | 202 | 103 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AB077438.1,NM_030887.2 transcripts.
Set Membership: |
|

Jdp2
Genomic Map