Assay Details
| Gene Symbol: Tmem63c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172583.2 | NP_766171.1 | 18 - 19 | 1835 | 93 | |
| GenBank mRNA | AK030062.1 | - | 18 - 19 | 1827 | 93 | |
| AK034410.1 | - | 17 - 18 | 1758 | 93 | ||
| AK043536.1 | - | 17 - 18 | 1756 | 93 | ||
| BC056936.1 | - | 18 - 19 | 1835 | 5695989 | 93 | |
| BC057088.1 | - | 18 - 19 | 1835 | 5695989 | 93 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tmem63c
Genomic Map