Assay Details
| Gene Symbol: Tmem63c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172583.2 | NP_766171.1 | 21 - 22 | 2172 | 85 | |
| GenBank mRNA | AK030062.1 | - | 21 - 22 | 2164 | 85 | |
| AK034410.1 | - | 20 - 21 | 2095 | 85 | ||
| AK043536.1 | - | 20 - 21 | 2094 | 85 | ||
| BC056936.1 | - | 21 - 22 | 2172 | 5695989 | 85 | |
| BC057088.1 | - | 21 - 22 | 2172 | 5695989 | 85 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tmem63c
Genomic Map