| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1200006327.mm.1 |
NM_172583.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200006327.mm.1 |
NM_172583.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200006327.mm.1 |
TC1200000866.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1200006328.mm.1 |
NM_172583.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200006328.mm.1 |
NM_172583.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200006328.mm.1 |
TC1200000866.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1200006324.mm.1 |
NM_172583.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200006324.mm.1 |
NM_172583.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200006324.mm.1 |
TC1200000866.mm.1 |
MTA TC |