Assay Details
| Gene Symbol: Hnf1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001291268.1 | NP_001278197.1 | 2 - 3 | 725 | 106 | |
| GenBank mRNA | AB008175.1 | - | 2 - 3 | 545 | 106 | |
| AB008177.1 | - | 2 - 3 | 545 | 106 | ||
| AK019258.1 | - | 2 - 3 | 357 | 106 | ||
More Information
Set Membership: |
|

Hnf1b
Genomic Map