Assay Details
| Gene Symbol: Rffl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007465.3 | NP_001007466.1 | 5 - 6 | 1097 | 112 | |
| NM_001164569.1 | NP_001158041.1 | 6 - 7 | 1241 | 112 | ||
| NM_001164570.1 | NP_001158042.1 | 6 - 7 | 1183 | 112 | ||
| NM_001164571.1 | NP_001158043.1 | 4 - 5 | 1016 | 112 | ||
| NM_026097.3 | NP_080373.1 | 4 - 5 | 1124 | 112 | ||
| GenBank mRNA | AF434814.1 | - | 3 - 4 | 802 | 112 | |
| AK006757.1 | - | 4 - 5 | 1112 | 112 | ||
| AK007189.1 | - | 4 - 5 | 1112 | 112 | ||
| AK028095.1 | - | 4 - 5 | 1113 | 112 | ||
| AK128983.1 | - | 6 - 7 | 1184 | 112 | ||
| AK150213.1 | - | 5 - 6 | 1030 | 112 | ||
| AK150541.1 | - | 6 - 7 | 1203 | 112 | ||
| AK159442.1 | - | 5 - 6 | 1056 | 112 | ||
| AK169393.1 | - | 4 - 5 | 957 | 112 | ||
| AK169988.1 | - | 5 - 6 | 114 | |||
| BC118517.1 | - | 4 - 5 | 899 | 40105713 | 112 | |
More Information
Set Membership: |
|

Rffl
Genomic Map