Assay Details
| Gene Symbol: Dynll2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001168471.1 | NP_001161943.1 | 1 - 2 | 86 | 58 | |
| GenBank mRNA | AK006876.1 | - | 1 - 2 | 68 | 58 | |
| BC049621.1 | - | 65 | 58 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001168471.1 transcripts.
Set Membership: |
|

Dynll2
Genomic Map