Assay Details
| Gene Symbol: Pde1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285890.1 | NP_001272819.1 | 2 - 3 | 556 | 72 | |
| NM_008800.2 | NP_032826.1 | 3 - 4 | 573 | 72 | ||
| GenBank mRNA | AK004772.1 | - | 2 - 3 | 163 | 72 | |
| BC058531.1 | - | 3 - 4 | 573 | 6826674 | 72 | |
| CV784321.1 | - | 2 - 3 | 612 | 72 | ||
| L01695.1 | - | 3 - 4 | 316 | 72 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pde1b
Genomic Map