Assay Details
| Gene Symbol: Mtch1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019880.3 | NP_063933.1 | 6 - 7 | 817 | 101 | |
| GenBank mRNA | AF176007.2 | - | 6 - 7 | 818 | 101 | |
| AF192558.2 | - | 6 - 7 | 818 | 101 | ||
| AK154010.1 | - | 6 - 7 | 673 | 101 | ||
| AK178745.1 | - | 1 - 2 | 53 | 101 | ||
| AK178762.1 | - | 1 - 2 | 52 | 100 | ||
| AK183124.1 | - | 1 - 2 | 53 | 101 | ||
| AK195241.1 | - | 1 - 2 | 53 | 101 | ||
| AK199429.1 | - | 1 - 2 | 53 | 101 | ||
| AK201694.1 | - | 1 - 2 | 53 | 101 | ||
| AK207873.1 | - | 1 - 2 | 53 | 101 | ||
| AK214576.1 | - | 1 - 2 | 53 | 101 | ||
| BC023135.1 | - | 6 - 7 | 660 | 3669288 | 101 | |
| BC026436.1 | - | 2 - 3 | 102 | 101 | ||
| BC027274.1 | - | 6 - 7 | 666 | 3156729 | 101 | |
More Information
Set Membership: |
|

Mtch1
Genomic Map