Assay Details
| Gene Symbol: Mtch1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF176007.2 | - | 8 - 9 | 973 | 81 | |
| AK154010.1 | - | 8 - 9 | 828 | 81 | ||
| AK207873.1 | - | 3 - 4 | 208 | 81 | ||
| BC026436.1 | - | 4 - 5 | 257 | 81 | ||
| BC027274.1 | - | 8 - 9 | 821 | 3156729 | 81 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mtch1
Genomic Map