Assay Details
| Gene Symbol: Acvrl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277255.1 | NP_001264184.1 | 3 - 4 | 261 | 112 | |
| GenBank mRNA | Z31664.1 | - | 2 - 3 | 206 | 111 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001277255.1, Z31664.1 transcripts.
Set Membership: |
|

Acvrl1
Genomic Map