Assay Details
| Gene Symbol: Acvrl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277258.1 | NP_001264187.1 | 2 - 3 | 202 | 81 | |
| GenBank mRNA | BC015083.1 | - | 1 - 2 | 152 | 3980151 | 81 |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC015083.1, NM_001277258.1 transcripts.
Set Membership: |
|

Acvrl1
Genomic Map